siRNA Details
Gene Symbol: FAM127B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078172.1 | NP_001071640.1 | 1 | 857 | ||
NM_001134321.1 | NP_001127793.1 | 2 | 309 | |||
GenBank mRNA | AL117556.1 | - | 1 | 796 | ||
BC000393.2 | - | 1 | 830 | 2821721 | ||
BC008096.1 | - | 1 | 615 | |||
BC019300.1 | - | 1 | 830 | 2821721 | ||
BC071991.1 | - | 2 | 220 | |||
CN367136.1 | - | 2 | 263 | |||
CR457123.1 | - | 1 | 173 | |||
CR542228.1 | - | 1 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
