siRNA Details
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161576.2 | NP_001155048.2 | 8 | 1944 | ||
NM_001161577.1 | NP_001155049.1 | 5 | 837 | |||
NM_015589.5 | NP_056404.4 | 9 | 2208 | |||
GenBank mRNA | AB028976.1 | - | 7 | 1275 | ||
AK024652.1 | - | 9 | 1901 | |||
AK025923.1 | - | 7 | 1089 | |||
AL117523.1 | - | 1 | 158 | |||
BC021060.1 | - | 5 | 754 | |||
BC021726.2 | - | 5 | 792 | |||
BC121173.1 | - | 8 | 1641 | 40122465 | ||
BC121174.1 | - | 8 | 1641 | 40122467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
