siRNA Details
Gene Symbol: FLJ33360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028351.1 | 2 | 451 | |||
GenBank mRNA | AK090679.1 | - | 2 | 451 | ||
BC132707.1 | - | 2 | 439 | 40146729 | ||
BC132709.1 | - | 2 | 439 | 40146731 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
