siRNA Details
Gene Symbol: LINC00469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027146.1 | 2 | 123 | |||
GenBank mRNA | AK097638.1 | - | 2 | 123 | ||
BC101213.1 | - | 2 | 106 | |||
BC101214.1 | - | 2 | 106 | |||
BC101215.1 | - | 2 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
