siRNA Details
Gene Symbol: RPP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017793.2 | NP_060263.2 | 1 | 865 | ||
GenBank mRNA | AK000381.1 | - | 1 | 864 | ||
AY034074.1 | - | 1 | 44 | |||
BC002497.2 | - | 1 | 183 | 3050784 | ||
BC007270.1 | - | 1 | 195 | 3160319 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133982, BC083064, BC016085, AK082822, AK140177.
Gene Ontology Categories:
