siRNA Details
Gene Symbol: RPP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017793.2 | NP_060263.2 | 1 | 970 | ||
GenBank mRNA | AK000381.1 | - | 1 | 969 | ||
AY034074.1 | - | 1 | 149 | |||
BC002497.2 | - | 1 | 288 | 3050784 | ||
BC007270.1 | - | 1 | 300 | 3160319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
