siRNA Details
Gene Symbol: RPRML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203400.4 | NP_981945.1 | 1 | 456 | ||
GenBank mRNA | BC033942.1 | - | 1 | 461 | 5274621 | |
BC108690.1 | - | 1 | 431 | 6140781 |

More Information
Other Species Targeted

