siRNA Details
Gene Symbol: RPRML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203400.4 | NP_981945.1 | 1 | 578 | ||
GenBank mRNA | BC033942.1 | - | 1 | 583 | 5274621 | |
BC108690.1 | - | 1 | 553 | 6140781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
