siRNA Details
Gene Symbol: PRR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175922.3 | NP_787118.2 | 1 | 404 | ||
GenBank mRNA | BC034775.1 | - | 1 | 404 | 5177440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
