siRNA Details
Gene Symbol: WWC2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024008.1 | 1 | 1029 | |||
GenBank mRNA | AK054839.1 | - | 1 | 1029 | ||
BC126477.1 | - | 1 | 575 | |||
BC126479.1 | - | 1 | 575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
