siRNA Details
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164093.1 | NP_001157565.1 | 7 | 1902 | ||
NM_001164094.1 | NP_001157566.1 | 8 | 1767 | |||
NM_001164095.2 | NP_001157567.1 | 8 | 1838 | |||
NM_016319.3 | NP_057403.1 | 8 | 1740 | |||
XM_005253694.2 | XP_005253751.1 | 8 | 1665 | |||
GenBank mRNA | AF193844.1 | - | 7 | 1528 | ||
AF210052.1 | - | 8 | 1669 | |||
AK001318.1 | - | 8 | 1652 | |||
AK021721.1 | - | 8 | 1705 | |||
AK022488.1 | - | 8 | 1647 | |||
AK128792.1 | - | 6 | 1437 | |||
BC011789.2 | - | 8 | 1628 | 4025790 | ||
BC093015.1 | - | 8 | 1293 | 30344094 |
