siRNA Details
Gene Symbol: CITED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133467.2 | NP_597724.1 | 1 | 449 | ||
GenBank mRNA | BC035496.2 | - | 1 | 451 | 3622207 | |
BC052559.1 | - | 1 | 449 | 6250908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
