siRNA Details
Gene Symbol: CLDN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194284.2 | NP_919260.2 | 1 | 832 | ||
GenBank mRNA | AK123547.1 | - | 2 | 457 | ||
BC125148.1 | - | 1 | 812 | 40120346 | ||
BC125149.1 | - | 1 | 812 | 40120347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
