siRNA Details
Gene Symbol: LINC00174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026873.1 | 5 | 2930 | |||
GenBank mRNA | AK091213.1 | - | 1 | 1588 | ||
AK092625.1 | - | 1 | 707 | |||
AK124597.1 | - | 1 | 2051 | |||
BC020399.1 | - | 5 | 2932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
