siRNA Details
Gene Symbol: LINC01465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121682.1 | 1 | 115 | |||
GenBank mRNA | AK098456.1 | - | 1 | 116 | ||
BC136288.1 | - | 1 | 109 | 9020275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
