siRNA Details
Gene Symbol: GGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001560.2 | NP_001001560.1 | 15 | 2703 | ||
NM_001172687.1 | NP_001166158.1 | 17 | 2952 | |||
NM_001172688.1 | NP_001166159.1 | 18 | 3010 | |||
NM_013365.4 | NP_037497.1 | 17 | 2964 | |||
XM_005261517.3 | XP_005261574.1 | 18 | 3189 | |||
XM_005261518.1 | XP_005261575.1 | 17 | 2649 | |||
XM_005261520.1 | XP_005261577.1 | 17 | 2855 | |||
XM_005261522.1 | XP_005261579.1 | 12 | 2206 | |||
XM_006724229.1 | XP_006724292.1 | 18 | 2897 | |||
XM_011530122.1 | XP_011528424.1 | 18 | 3122 | |||
XM_011530123.1 | XP_011528425.1 | 17 | 2700 | |||
XM_011530124.1 | XP_011528426.1 | 16 | 2928 | |||
XM_017028760.1 | XP_016884249.1 | 18 | 3138 | |||
XM_017028761.1 | XP_016884250.1 | 16 | 4305 | |||
XM_017028762.1 | XP_016884251.1 | 16 | 2877 | |||
XM_017028763.1 | XP_016884252.1 | 18 | 2824 | |||
XM_017028764.1 | XP_016884253.1 | 15 | 2594 | |||
XM_017028765.1 | XP_016884254.1 | 11 | 2139 | |||
GenBank mRNA | AF190862.1 | - | 17 | 2592 | ||
AK075256.1 | - | 17 | 2942 | |||
AK092417.1 | - | 6 | 1611 | |||
AK095427.1 | - | 10 | 1854 | |||
AK122898.1 | - | 18 | 3010 | |||
AL110219.1 | - | 16 | 2488 | |||
BC000538.2 | - | 8 | 1757 | |||
BC044629.1 | - | 15 | 2332 | 5796877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
