siRNA Details
Gene Symbol: ZNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282795.1 | NP_001269724.1 | 3, 4 | 305 | ||
NM_001282796.1 | NP_001269725.1 | 3, 4 | 272 | |||
NM_003416.3 | NP_003407.1 | 3, 4 | 272 | |||
XM_011517291.2 | XP_011515593.1 | 3, 4 | 319 | |||
XM_011517292.2 | XP_011515594.1 | 3, 4 | 653 | |||
XM_011517293.2 | XP_011515595.1 | 3, 4 | 652 | |||
XM_011517294.2 | XP_011515596.1 | 3, 4 | 431 | |||
XM_017013814.1 | XP_016869303.1 | 2, 3 | 618 | |||
XM_017013816.1 | XP_016869305.1 | 3, 4 | 341 | |||
XM_017013817.1 | XP_016869306.1 | 3, 4 | 263 | |||
GenBank mRNA | AK096025.1 | - | 3, 4 | 231 | ||
AK300001.1 | - | 3, 4 | 240 | |||
AK308002.1 | - | 3, 4 | 264 | |||
AK313745.1 | - | 3, 4 | 212 | |||
BC058923.1 | - | 3, 4 | 216 | 4516166 | ||
BX647998.1 | - | 3, 4 | 231 | |||
DC301751.1 | - | 3, 4 | 330 | |||
M29580.1 | - | 3, 4 | 373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
