siRNA Details
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 3 | 423 | ||
XM_005266089.2 | XP_005266146.2 | 3 | 443 | |||
XM_011518789.1 | XP_011517091.1 | 3 | 443 | |||
XM_017014812.1 | XP_016870301.1 | 3 | 443 | |||
GenBank mRNA | AK026081.1 | - | 3 | 287 | ||
AK315738.1 | - | 3 | 298 | |||
BC014130.1 | - | 3 | 319 | 4620371 | ||
BC014297.2 | - | 3 | 290 | 3958395 | ||
BC031100.1 | - | 3 | 365 | |||
BC035355.1 | - | 3 | 357 | |||
BI829307.1 | - | 3 | 390 | |||
CF132404.1 | - | 3 | 364 | |||
DB460097.1 | - | 3 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
