siRNA Details
Gene Symbol: ADRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000682.6 | NP_000673.2 | 1 | 1357 | ||
GenBank mRNA | BC133021.1 | - | 1 | 1357 | 40147043 | |
BC136537.1 | - | 1 | 1357 | 9020527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
