siRNA Details
Gene Symbol: C11orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271562.1 | NP_001258491.1 | 1 | 344 | ||
NM_019021.3 | NP_061894.2 | 1 | 344 | |||
GenBank mRNA | AB101209.1 | - | 1 | 333 | ||
AK000017.1 | - | 1 | 305 | |||
BC009041.1 | - | 1 | 273 | 3868744 | ||
BC071695.1 | - | 1 | 211 | 5416335 | ||
CR457222.1 | - | 1 | 125 | |||
HY013971.1 | - | 1 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
