siRNA Details
Gene Symbol: CLDN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020982.3 | NP_066192.1 | 1 | 2024 | ||
GenBank mRNA | AK091002.1 | - | 1 | 2024 | ||
BC051870.1 | - | 2 | 1689 | 6172607 | ||
BC065830.1 | - | 1 | 1352 | 30364848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
