siRNA Details
Gene Symbol: FAAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025161.5 | NP_079437.5 | 7 | 2439 | ||
NR_033338.1 | 7 | 2635 | ||||
XM_006722111.2 | XP_006722174.1 | 7 | 2020 | |||
XM_006722112.3 | XP_006722175.1 | 7 | 2585 | |||
XM_017025162.1 | XP_016880651.1 | 7 | 2460 | |||
XM_017025163.1 | XP_016880652.1 | 7 | 2356 | |||
GenBank mRNA | AK024644.1 | - | 6 | 866 | ||
AK025828.1 | - | 3 | 700 | |||
AK054713.1 | - | 3 | 1783 | |||
AK225370.1 | - | 3 | 700 | |||
AL834374.1 | - | 3 | 924 | |||
BC001724.1 | - | 2 | 198 | |||
BC008883.2 | - | 7 | 2608 | |||
BC021968.2 | - | 3 | 511 | |||
BC117139.1 | - | 5 | 1959 | 40125690 | ||
BC117141.1 | - | 5 | 1959 | 40125692 | ||
BC143861.1 | - | 5 | 1931 | |||
DQ989324.1 | - | 7 | 2421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
