siRNA Details
Gene Symbol: FAAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025161.5 | NP_079437.5 | 9 | 2616 | ||
NR_033338.1 | 9 | 2812 | ||||
XM_006722111.2 | XP_006722174.1 | 9 | 2197 | |||
XM_006722112.3 | XP_006722175.1 | 9 | 2762 | |||
XM_017025162.1 | XP_016880651.1 | 9 | 2637 | |||
XM_017025163.1 | XP_016880652.1 | 9 | 2533 | |||
GenBank mRNA | AK024644.1 | - | 8 | 1043 | ||
AK025828.1 | - | 5 | 877 | |||
AK054713.1 | - | 5 | 1960 | |||
AK225370.1 | - | 5 | 877 | |||
AL834374.1 | - | 5 | 1101 | |||
BC001724.1 | - | 4 | 375 | |||
BC008883.2 | - | 9 | 2785 | |||
BC021968.2 | - | 5 | 688 | |||
BC117139.1 | - | 7 | 2136 | 40125690 | ||
BC117141.1 | - | 7 | 2136 | 40125692 | ||
BC143861.1 | - | 7 | 2108 | |||
DQ989324.1 | - | 9 | 2598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
