siRNA Details
Gene Symbol: ZNF639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303425.1 | NP_001290354.1 | 7 | 882 | ||
NM_001303426.1 | NP_001290355.1 | 6 | 838 | |||
NM_016331.2 | NP_057415.1 | 7 | 900 | |||
XM_017006550.1 | XP_016862039.1 | 7 | 1545 | |||
XM_017006551.1 | XP_016862040.1 | 7 | 1200 | |||
XM_017006552.1 | XP_016862041.1 | 7 | 891 | |||
XM_017006553.1 | XP_016862042.1 | 7 | 1308 | |||
GenBank mRNA | AB097862.1 | - | 7 | 900 | ||
AF003924.1 | - | 7 | 900 | |||
BC020500.1 | - | 6 | 673 | 3912907 | ||
BC026181.1 | - | 6 | 803 | 4794621 | ||
DQ323613.1 | - | 3 | 309 |
