siRNA Details
Gene Symbol: MIR137HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092728.1 | - | 1 | 530 | ||
AK309618.1 | - | 2 | 539 | |||
AK311400.1 | - | 2 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
