siRNA Details
Gene Symbol: LOC150776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026922.1 | 17 | 2265 | |||
GenBank mRNA | AK123346.1 | - | 2 | 654 | ||
BC093080.1 | - | 17 | 1988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
