siRNA Details
Gene Symbol: C1QTNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207419.3 | NP_997302.2 | 3 | 467 | ||
XM_011522483.2 | XP_011520785.1 | 3 | 550 | |||
GenBank mRNA | AY358832.1 | - | 3 | 467 | ||
FJ422561.1 | - | 2 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
