siRNA Details
Gene Symbol: OAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178507.2 | NP_848602.1 | 3, 4 | 792 | ||
GenBank mRNA | AY459296.1 | - | 3, 4 | 551 | ||
BC047726.1 | - | 3, 4 | 792 | 5763917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
