siRNA Details
Gene Symbol: ADARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018702.3 | NP_061172.1 | 10 | 2405 | ||
GenBank mRNA | AF034837.1 | - | 10 | 2413 | ||
AK055107.1 | - | 3 | 610 | |||
AK057763.1 | - | 3 | 308 | |||
AK289895.1 | - | 3 | 642 | |||
AK307451.1 | - | 3 | 475 | |||
AK308579.1 | - | 4 | 380 | |||
BC047443.1 | - | 3 | 279 | |||
BC137477.1 | - | 10 | 2226 | 9021477 | ||
BC139725.1 | - | 3 | 286 | |||
BC140852.1 | - | 10 | 2226 | 9021720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
