siRNA Details
Gene Symbol: S100A7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176823.3 | NP_789793.1 | 3 | 213 | ||
GenBank mRNA | AY189118.1 | - | 3 | 213 | ||
AY189119.1 | - | 2 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
