siRNA Details
Gene Symbol: PRSS57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308209.1 | NP_001295138.1 | 3 | 386 | ||
NM_214710.4 | NP_999875.1 | 3 | 389 | |||
GenBank mRNA | AY358594.1 | - | 3 | 426 | ||
BC137161.1 | - | 3 | 391 | 9021158 | ||
BC144505.1 | - | 3 | 388 | 9053032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
