siRNA Details
Gene Symbol: PRR23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013650.2 | NP_001013672.1 | 1 | 693 | ||
GenBank mRNA | AK127998.1 | - | 1 | 693 | ||
BC137145.1 | - | 1 | 681 | 9021142 | ||
BC137146.1 | - | 1 | 679 | 9021143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
