siRNA Details
Gene Symbol: B3GALT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026542.1 | 1, 2 | 536 | |||
NR_026543.1 | 1, 2 | 536 | ||||
GenBank mRNA | AF426266.1 | - | 1, 2 | 155 | ||
AF426267.1 | - | 1, 2 | 155 | |||
BC080530.1 | - | 1, 2 | 535 | |||
BC119737.2 | - | 1, 2 | 129 | |||
BC119738.2 | - | 1, 2 | 129 | |||
BG751912.1 | - | 1, 2 | 535 | |||
DA218783.1 | - | 1, 2 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
