siRNA Details
Gene Symbol: KRTAP8-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175857.3 | NP_787053.1 | 1 | 202 | ||
GenBank mRNA | AB096961.1 | - | 1 | 200 | ||
AJ457064.1 | - | 1 | 202 | |||
BC130397.1 | - | 1 | 193 | 40146428 | ||
BC133033.1 | - | 1 | 193 | 40147055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
