siRNA Details
Gene Symbol: ASMTL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026710.1 | 4 | 1080 | |||
NR_026711.1 | 4 | 1080 | ||||
GenBank mRNA | AK023392.1 | - | 4 | 513 | ||
BC093994.2 | - | 3 | 423 | |||
BC143527.1 | - | 3 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
