siRNA Details
Gene Symbol: C9orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152569.2 | NP_689782.2 | 1 | 510 | ||
GenBank mRNA | AK055720.1 | - | 1 | 358 | ||
BC111423.1 | - | 1 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
