siRNA Details
Gene Symbol: C2orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206895.1 | NP_996778.1 | 1, 2 | 137 | ||
XM_017004080.1 | XP_016859569.1 | 2, 3 | 1167 | |||
XM_017004081.1 | XP_016859570.1 | 2, 3 | 905 | |||
XM_017004082.1 | XP_016859571.1 | 3, 4 | 1225 | |||
XM_017004083.1 | XP_016859572.1 | 2, 3 | 937 | |||
XM_017004084.1 | XP_016859573.1 | 2, 3 | 200 | |||
GenBank mRNA | AY358535.1 | - | 1, 2 | 137 | ||
BC130307.1 | - | 1, 2 | 135 | 40146338 | ||
BC130309.1 | - | 1, 2 | 135 | 40146340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
