siRNA Details
Gene Symbol: C2orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206895.1 | NP_996778.1 | 2 | 167 | ||
XM_017004080.1 | XP_016859569.1 | 3 | 1197 | |||
XM_017004081.1 | XP_016859570.1 | 3 | 935 | |||
XM_017004082.1 | XP_016859571.1 | 4 | 1255 | |||
XM_017004083.1 | XP_016859572.1 | 3 | 967 | |||
XM_017004084.1 | XP_016859573.1 | 3 | 230 | |||
GenBank mRNA | AY358535.1 | - | 2 | 167 | ||
BC035093.2 | - | 1 | 156 | |||
BC130307.1 | - | 2 | 165 | 40146338 | ||
BC130309.1 | - | 2 | 165 | 40146340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
