siRNA Details
Gene Symbol: HS3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006040.2 | NP_006031.2 | 2 | 1723 | ||
GenBank mRNA | AF105378.2 | - | 2 | 1720 | ||
AL834283.1 | - | 2 | 970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
