siRNA Details
Gene Symbol: KLHL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153270.2 | NP_695002.1 | 1 | 720 | ||
GenBank mRNA | AK092279.1 | - | 1 | 663 | ||
BC109031.2 | - | 1 | 632 | 40027951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
