siRNA Details
Gene Symbol: CECR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031890.3 | NP_114096.1 | 1 | 1172 | ||
XM_011546124.2 | XP_011544426.1 | 1 | 1199 | |||
GenBank mRNA | AF307451.1 | - | 1 | 1102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
