siRNA Details
Gene Symbol: LDOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012317.3 | NP_036449.1 | 1 | 242 | ||
GenBank mRNA | AB019527.1 | - | 1 | 153 | ||
BC003104.1 | - | 1 | 127 | 3503444 | ||
CR457088.1 | - | 1 | 49 | |||
HQ343285.1 | - | 1 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
