siRNA Details
Gene Symbol: LINC00482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038080.1 | 4 | 559 | |||
GenBank mRNA | AK096740.1 | - | 4 | 559 | ||
BC108932.1 | - | 3 | 507 | 40025762 | ||
BC108933.1 | - | 3 | 507 | 40025767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
