siRNA Details
Gene Symbol: SIAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198849.2 | NP_942146.2 | 2 | 3317 | ||
GenBank mRNA | AK125411.1 | - | 1 | 1471 | ||
BC041372.2 | - | 2 | 3319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
