siRNA Details
Gene Symbol: EP400NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003290.2 | 4 | 330 | |||
GenBank mRNA | AK090610.1 | - | 5 | 1902 | ||
AK091234.1 | - | 4 | 1457 | |||
AK097720.1 | - | 5 | 1022 | |||
AK126212.1 | - | 4 | 1578 | |||
AK289710.1 | - | 6 | 1533 | |||
AL117637.1 | - | 2 | 1157 | |||
BC066974.1 | - | 6 | 1456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
