siRNA Details
Gene Symbol: C7orf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026865.1 | 1 | 1168 | |||
GenBank mRNA | AF063598.1 | - | 1 | 935 | ||
AF439977.1 | - | 1 | 1168 | |||
AK056371.1 | - | 1 | 1084 | |||
BC031037.1 | - | 1 | 1053 | |||
BC070228.1 | - | 2 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
