siRNA Details
Gene Symbol: LENG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301782.1 | NP_001288711.1 | 1 | 1088 | ||
NM_198988.1 | NP_945339.2 | 1 | 1091 | |||
GenBank mRNA | AF211976.1 | - | 1 | 220 | ||
BC015921.1 | - | 1 | 1091 | 4551151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
