siRNA Details
Gene Symbol: FOXI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207426.2 | NP_997309.2 | 2 | 841 | ||
GenBank mRNA | AK128865.1 | - | 2 | 427 | ||
BC137296.1 | - | 2 | 383 | |||
BC137297.1 | - | 2 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
