siRNA Details
Gene Symbol: KRTAP19-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303120.1 | NP_001290049.1 | 1 | 44 | ||
NM_181612.3 | NP_853643.1 | 1 | 44 | |||
GenBank mRNA | AB096947.1 | - | 1 | 42 | ||
AB096948.1 | - | 1 | 42 | |||
BC101813.1 | - | 1 | 44 | |||
BC101815.1 | - | 1 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
