siRNA Details
Gene Symbol: TDRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153046.2 | NP_694591.2 | 36 | 4394 | ||
XM_005267309.3 | XP_005267366.1 | 35 | 4375 | |||
XM_006720019.3 | XP_006720082.1 | 37 | 4604 | |||
XM_006720020.3 | XP_006720083.1 | 35 | 4450 | |||
XM_011536397.2 | XP_011534699.1 | 35 | 4426 | |||
XM_011536398.2 | XP_011534700.1 | 36 | 4336 | |||
XM_011536400.2 | XP_011534702.1 | 33 | 4270 | |||
XM_011536401.2 | XP_011534703.1 | 36 | 4269 | |||
XM_011536402.2 | XP_011534704.1 | 29 | 3367 | |||
XM_017020955.1 | XP_016876444.1 | 35 | 4198 | |||
GenBank mRNA | AK093483.1 | - | 9 | 1324 | ||
AK097429.1 | - | 20 | 2464 | |||
AK097699.1 | - | 13 | 1866 | |||
AK125978.1 | - | 26 | 2970 | |||
AL833915.1 | - | 33 | 3784 | |||
BC016796.1 | - | 4 | 587 | |||
BC042804.1 | - | 2 | 581 | |||
U56244.1 | - | 23 | 2260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
